Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178036.4 | NP_828875.1 | 5 | 510 | ||
GenBank mRNA | AF435737.1 | - | 5 | 510 | ||
AK020338.1 | - | 4 | 258 | |||
BC119103.1 | - | 5 | 478 | 8733852 | ||
BC120657.1 | - | 5 | 478 | 40129580 |