Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181548.2 | NP_853526.1 | 2 | 360 | ||
GenBank mRNA | AB093573.1 | - | 2 | 360 | ||
AK141212.1 | - | 2 | 254 | |||
BC119277.1 | - | 2 | 276 | 8734026 | ||
BC119281.1 | - | 2 | 276 | 8734030 |