Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181820.2 | NP_861541.2 | 14, 15 | 2080 | ||
GenBank mRNA | AK050105.1 | - | 13, 14 | 1975 | ||
AY236493.1 | - | 14, 15 | 2043 | |||
AY263162.1 | - | 14, 15 | 2051 | |||
BC022758.1 | - | 6, 7 | 770 | |||
BC040466.1 | - | 14, 15 | 1992 | |||
CN719678.1 | - | 3, 4 | 274 |