Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004157.2 | NP_001004157.2 | 8 | 1298 | ||
GenBank mRNA | AK133477.1 | - | 8 | 1281 | ||
AK220215.1 | - | 8 | 1282 | |||
BC057600.1 | - | 7 | 1205 | |||
BC080288.1 | - | 8 | 1298 | 6411836 |