Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081393.1 | NP_001074862.1 | 17 | 2598 | ||
GenBank mRNA | BC158097.1 | - | 16 | 2587 | 9088178 |