Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027928.1 | NP_082204.1 | 3 | 963 | ||
GenBank mRNA | AK004401.1 | - | 1 | 245 |