Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177505.1 | NP_001170976.1 | 3 | 674 | ||
GenBank mRNA | BC042569.1 | - | 4 | 955 | ||
BC119591.1 | - | 3 | 674 | 40058904 |