Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198680.2 | NP_941373.1 | 8 | 843 | ||
GenBank mRNA | AK003220.1 | - | 8 | 844 | ||
AY144685.1 | - | 7 | 803 | |||
AY144686.1 | - | 7 | 803 | |||
AY367774.1 | - | 7 | 787 | |||
BC099584.1 | - | 8 | 840 | 30291007 |