Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201376.1 | NP_958764.1 | 4 | 410 | ||
GenBank mRNA | AY367776.1 | - | 4 | 410 | ||
BC138290.1 | - | 4 | 436 | 8861309 | ||
BC138291.1 | - | 4 | 436 | 8861310 |