Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001183.1 | NP_001001183.1 | 3 | 1220 | ||
GenBank mRNA | BC019551.1 | - | 3 | 1234 | ||
BC043726.1 | - | 3 | 1220 | |||
BC054438.1 | - | 3 | 1220 | 6390142 |