Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011804.2 | NP_035934.1 | 3 | 600 | ||
GenBank mRNA | AF084524.1 | - | 3 | 600 | ||
AK005388.1 | - | 3 | 621 | |||
AK010947.1 | - | 3 | 617 | |||
AK085844.1 | - | 3 | 412 | |||
AK144101.1 | - | 3 | 619 | |||
AK150006.1 | - | 4 | 726 | |||
AK189906.1 | - | 3 | 311 | |||
AK190952.1 | - | 3 | 309 | |||
AK191906.1 | - | 3 | 309 | |||
AK193253.1 | - | 3 | 315 | |||
AK200846.1 | - | 3 | 309 | |||
AK204882.1 | - | 3 | 309 | |||
AK204927.1 | - | 2 | 216 | |||
AK219109.1 | - | 3 | 321 | |||
BC027426.1 | - | 3 | 600 | 3670649 |