Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm10665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167160.1 | NP_001160632.1 | 1 | 255 |
Gene Symbol: Gm10670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167161.1 | NP_001160633.1 | 1 | 255 | ||
GenBank mRNA | BC107204.1 | - | 1 | 254 |
Gene Symbol: Gm4207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC119491.1 | - | 254 |
Gene Symbol: Vmn1r-ps79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC115870.1 | - | 1 | 1314 | 40062110 | |
BC119492.1 | - | 1 | 254 |