Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm4207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC107205.1 | - | 1 | 397 | ||
BC119491.1 | - | 397 |
Gene Symbol: Gm10665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167160.1 | NP_001160632.1 | 1 | 398 |
Gene Symbol: Gm10666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167573.1 | NP_001161045.1 | 1 | 398 |
Gene Symbol: Gm10670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167161.1 | NP_001160633.1 | 1 | 398 | ||
GenBank mRNA | BC107204.1 | - | 1 | 397 |
Gene Symbol: Gm4175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167165.1 | NP_001160637.1 | 1 | 398 |
Gene Symbol: Gm4214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167164.1 | NP_001160636.1 | 1 | 398 |
Gene Symbol: Vmn1r-ps79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC115870.1 | - | 1 | 1454 | 40062110 | |
BC119492.1 | - | 1 | 394 |
Gene Symbol: Vmn1r114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166837.1 | NP_001160309.1 | 1 | 398 |
Gene Symbol: Vmn1r121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166741.1 | NP_001160213.1 | 1 | 398 |
Gene Symbol: Vmn1r126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166838.1 | NP_001160310.1 | 1 | 398 |
Gene Symbol: Vmn1r159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166758.1 | NP_001160230.1 | 1 | 398 |
Gene Symbol: Vmn1r93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207547.2 | NP_997430.2 | 1 | 398 |