Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siva1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013929.2 | NP_038957.2 | 2 | 297 | ||
GenBank mRNA | AF033113.1 | - | 2 | 158 | ||
AF033114.1 | - | 2 | 245 | |||
AK010562.1 | - | 2 | 223 | |||
AK017570.1 | - | 2 | 258 | |||
AK153115.1 | - | 2 | 207 | |||
AK169200.1 | - | 2 | 250 | |||
AK184852.1 | - | 2 | 102 | |||
AK209128.1 | - | 2 | 102 | |||
AK212226.1 | - | 2 | 102 | |||
BC012279.1 | - | 2 | 279 | 4013026 |