Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5430405H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134903.1 | - | 2 | 664 | ||
AK134962.1 | - | 2 | 898 | |||
BC042697.1 | - | 2 | 769 |
Gene Symbol: Tgif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291124.1 | NP_001278053.1 | 4 | 913 | ||
NM_173396.3 | NP_775572.1 | 3 | 1030 | |||
GenBank mRNA | AK017884.1 | - | 3 | 892 | ||
AK029490.1 | - | 3 | 960 | |||
AK157623.1 | - | 4 | 821 | |||
AK186967.1 | - | 1 | 39 | |||
BC138705.1 | - | 3 | 943 | 8861727 | ||
BC138706.1 | - | 3 | 945 | 8861728 | ||
BC158022.1 | - | 2 | 792 | 9007328 | ||
CF731285.1 | - | 3 | 531 |