Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037741.3 | NP_001032830.2 | 1 | 118 | ||
GenBank mRNA | AB072797.1 | - | 1 | 100 | ||
AF274027.1 | - | 1 | 94 | |||
BU961981.1 | - | 121 |