Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016963.2 | NP_058659.1 | 2 | 302 | ||
GenBank mRNA | AF177172.1 | - | 2 | 290 | ||
AF237630.1 | - | 2 | 216 | |||
AK017725.1 | - | 2 | 299 | |||
AK050372.1 | - | 2 | 304 | |||
AK082971.1 | - | 2 | 303 | |||
AK147017.1 | - | 2 | 303 | |||
AK167723.1 | - | 2 | 300 | |||
AK169218.1 | - | 2 | 303 | |||
BC082595.1 | - | 2 | 294 | 30645500 | ||
BY067594.1 | - | 2 | 303 |