Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016856.3 | NP_058552.1 | 10, 11 | 1634 | ||
GenBank mRNA | AF012822.1 | - | 10, 11 | 1620 | ||
AK140869.1 | - | 10, 11 | 1616 | |||
BC007163.1 | - | 8, 9 | 1292 | |||
BC013628.1 | - | 10, 11 | 1609 | 3497854 |