Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178608.4 | NP_848723.1 | 4 | 553 | ||
GenBank mRNA | AK035238.1 | - | 4 | 387 | ||
AK045056.1 | - | 4 | 375 | |||
AK053158.1 | - | 4 | 371 | |||
AK136916.1 | - | 4 | 332 | |||
AY562229.1 | - | 4 | 264 | |||
BC046826.1 | - | 4 | 324 | 6417744 | ||
CX239705.1 | - | 4 | 553 |