Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024499.1 | NP_078775.1 | 3 | 238 | ||
GenBank mRNA | AK039966.1 | - | 2 | 236 | ||
AK051005.1 | - | 3 | 240 | |||
AK079168.1 | - | 3 | 248 | |||
AK165557.1 | - | 3 | 265 | |||
AK179057.1 | - | 2 | 118 | |||
BC003836.1 | - | 3 | 238 | 3486148 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK003018.1 | - | 176 | Chr. - - [-] on Build |