Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdipt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026638.3 | NP_080914.1 | 1 | 877 | ||
GenBank mRNA | AK003041.1 | - | 1 | 428 | ||
AK076974.1 | - | 1 | 877 | |||
AK079226.1 | - | 1 | 354 | |||
AK089109.1 | - | 1 | 443 | |||
AK132286.1 | - | 1 | 446 | |||
BC021473.1 | - | 1 | 405 | 5343411 | ||
BC024413.1 | - | 1 | 217 | 4973307 | ||
BC032182.1 | - | 1 | 365 | 5252470 |