Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016880.2 | NP_058576.2 | 3 | 657 | ||
GenBank mRNA | AF020790.1 | - | 3 | 658 | ||
AK132593.1 | - | 3 | 675 | |||
BC100542.1 | - | 3 | 657 | 30295313 | ||
BC140965.1 | - | 3 | 609 | 9055577 |