Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016918.3 | NP_058614.1 | 7 | 853 | ||
GenBank mRNA | AF222786.1 | - | 6 | 503 | ||
AK088222.1 | - | 7 | 854 | |||
BC004571.1 | - | 7 | 594 | 3586340 | ||
BC049595.1 | - | 7 | 658 | 6774692 |