Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282072.1 | NP_001269001.1 | 4 | 791 | ||
NM_001282073.1 | NP_001269002.1 | 5 | 715 | |||
NM_001282076.1 | NP_001269005.1 | 5 | 748 | |||
NM_016874.3 | NP_058570.1 | 4 | 791 | |||
GenBank mRNA | AF102818.1 | - | 4 | 651 | ||
AI430175.1 | - | 3 | 273 | |||
AK082115.1 | - | 4 | 644 | |||
AK144333.1 | - | 4 | 560 | |||
AK146546.1 | - | 4 | 668 | |||
AK155836.1 | - | 4 | 757 | |||
AK158586.1 | - | 5 | 600 | |||
BC046399.1 | - | 5 | 693 | 5063995 | ||
FJ377318.1 | - | 5 | 608 | |||
FJ377319.1 | - | 4 | 636 |