Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282072.1 | NP_001269001.1 | 11 | 1703 | ||
NM_001282073.1 | NP_001269002.1 | 11 | 1457 | |||
NM_001282076.1 | NP_001269005.1 | 12 | 1660 | |||
NM_016874.3 | NP_058570.1 | 10 | 1533 | |||
GenBank mRNA | AA269946.1 | - | 1 | 51 | ||
AF102818.1 | - | 10 | 1393 | |||
AK082115.1 | - | 10 | 1386 | |||
AK144333.1 | - | 11 | 1472 | |||
AK146546.1 | - | 10 | 1410 | |||
AK155836.1 | - | 11 | 1669 | |||
AK158586.1 | - | 11 | 1342 | |||
BC046399.1 | - | 11 | 1435 | 5063995 | ||
CD353366.1 | - | 4 | 469 | |||
CF538574.1 | - | 2 | 235 | |||
FJ377318.1 | - | 12 | 1520 | |||
FJ377319.1 | - | 10 | 1378 |