Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Barhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164186.1 | NP_001157658.1 | 2 | 593 | ||
NM_019446.4 | NP_062319.1 | 1 | 876 | |||
GenBank mRNA | AB043980.1 | - | 1 | 891 | ||
AF264026.1 | - | 3 | 651 | |||
AJ237590.1 | - | 1 | 151 | |||
AK036312.1 | - | 1 | 327 | |||
BC055731.1 | - | 2 | 594 | 6413504 |