Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018810.2 | NP_061280.2 | 8 | 1422 | ||
GenBank mRNA | AB026986.1 | - | 8 | 1542 | ||
AF192785.1 | - | 8 | 1447 | |||
AK029500.1 | - | 8 | 1462 | |||
AK077060.1 | - | 8 | 1423 |