Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cts7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019539.3 | NP_062412.1 | 8 | 1161 | ||
GenBank mRNA | AF250837.1 | - | 7 | 948 | ||
AK017466.1 | - | 8 | 1185 | |||
AY014779.1 | - | 8 | 1021 | |||
BC050198.1 | - | 7 | 1257 | |||
BC064740.1 | - | 8 | 1161 | 30018572 |