Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cts7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019539.3 | NP_062412.1 | 6 | 864 | ||
GenBank mRNA | AF250837.1 | - | 5 | 651 | ||
AK017466.1 | - | 6 | 888 | |||
AY014779.1 | - | 6 | 724 | |||
BC050198.1 | - | 5 | 960 | |||
BC064740.1 | - | 6 | 864 | 30018572 |