Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085472.2 | NP_001078941.2 | 10 | 1040 | ||
NM_001085473.2 | NP_001078942.2 | 16 | 3365 | |||
NM_001242605.1 | NP_001229534.1 | 9 | 1104 | |||
NM_001242606.1 | NP_001229535.1 | 9 | 1065 | |||
NM_019567.3 | NP_062513.3 | 10 | 1169 | |||
NM_023190.3 | NP_075679.2 | 16 | 3401 | |||
GenBank mRNA | AF124725.1 | - | 10 | 1100 | ||
AF124729.1 | - | 9 | 1035 | |||
AF168782.1 | - | 16 | 3402 | |||
AK012099.1 | - | 10 | 1033 | |||
AK030449.1 | - | 16 | 3095 | |||
AK122342.1 | - | 16 | 3036 | |||
BC052755.1 | - | 9 | 1025 | |||
BC075691.1 | - | 16 | 3050 | |||
BC094217.1 | - | 9 | 1047 |