Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019868.3 | NP_063921.1 | 4 | 742 | ||
GenBank mRNA | AK077716.1 | - | 4 | 743 | ||
AK164115.1 | - | 3 | 2663 | |||
AK168116.1 | - | 3 | 725 | |||
AK169104.1 | - | 3 | 706 | |||
BC005461.1 | - | 3 | 724 | 3496629 |