Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019657.4 | NP_062631.1 | 3 | 284 | ||
GenBank mRNA | AF064635.1 | - | 3 | 263 | ||
AK076088.1 | - | 3 | 270 | |||
AK078841.1 | - | 3 | 285 | |||
AK081869.1 | - | 3 | 278 | |||
AK082715.1 | - | 3 | 278 | |||
AK088521.1 | - | 3 | 278 | |||
AK150849.1 | - | 3 | 279 | |||
AK152489.1 | - | 3 | 279 | |||
AK166027.1 | - | 3 | 278 | |||
AK167908.1 | - | 3 | 278 | |||
BC010749.1 | - | 3 | 260 | |||
BC090659.1 | - | 3 | 254 | 30675153 |