Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019701.2 | NP_062675.2 | 16 | 1683 | ||
GenBank mRNA | AA240610.1 | - | 2 | 85 | ||
AF124847.1 | - | 16 | 1676 | |||
AK146876.1 | - | 15 | 1746 | |||
BC019983.1 | - | 16 | 1709 | 4188062 |