Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019701.2 | NP_062675.2 | 18 | 1905 | ||
GenBank mRNA | AA240610.1 | - | 4 | 307 | ||
AF124847.1 | - | 18 | 1898 | |||
AK146876.1 | - | 17 | 1967 |