Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029565.3 | NP_083841.4 | 7 | 862 | ||
GenBank mRNA | AF116911.1 | - | 7 | 813 | ||
AK003252.1 | - | 7 | 776 | |||
AK014184.1 | - | 7 | 648 | |||
AK050162.1 | - | 7 | 780 | |||
AK145644.1 | - | 7 | 783 | |||
AK159761.1 | - | 7 | 790 | |||
AK166772.1 | - | 7 | 777 | |||
AK167058.1 | - | 7 | 776 | |||
AK177231.1 | - | 3 | 195 | |||
AX775905.1 | - | 7 | 797 | |||
BC002164.1 | - | 1 | 41 | |||
BC014732.1 | - | 7 | 766 | 4235248 | ||
BC018379.1 | - | 7 | 784 | 4242583 | ||
BC045145.1 | - | 7 | 800 | 6475115 | ||
BC058273.1 | - | 7 | 777 | 6494379 |