Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019793.3 | NP_062767.3 | 6 | 844 | ||
GenBank mRNA | AB009021.1 | - | 6 | 626 | ||
AB072450.1 | - | 6 | 654 | |||
AF133427.1 | - | 6 | 638 | |||
AF242591.1 | - | 6 | 837 | |||
AK006798.1 | - | 6 | 720 | |||
AK053325.1 | - | 6 | 695 | |||
AK089494.1 | - | 676 | ||||
AK141951.1 | - | 6 | 702 | |||
AK146130.1 | - | 6 | 832 | |||
AK152175.1 | - | 6 | 729 | |||
AK154919.1 | - | 676 | ||||
AK159324.1 | - | 6 | 815 | |||
AK159418.1 | - | 6 | 788 | |||
AK159621.1 | - | 6 | 799 | |||
AK159914.1 | - | 6 | 777 | |||
AK168231.1 | - | 6 | 832 | |||
AK169068.1 | - | 6 | 815 | |||
AK178223.1 | - | 4 | 324 | |||
AK180027.1 | - | 4 | 324 | |||
AK183645.1 | - | 4 | 325 | |||
AK187762.1 | - | 3 | 311 | |||
AK207599.1 | - | 3 | 322 | |||
AK210479.1 | - | 3 | 291 | |||
AK220124.1 | - | 4 | 322 | |||
BC043072.1 | - | 6 | 822 | 6404165 |