Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291205.1 | NP_001278134.1 | 3 | 301 | ||
NM_001291206.1 | NP_001278135.1 | 3 | 652 | |||
NM_001291207.1 | NP_001278136.1 | 2 | 197 | |||
NM_001291208.1 | NP_001278137.1 | 2 | 546 | |||
NM_001291209.1 | NP_001278138.1 | 3 | 303 | |||
NM_026446.4 | NP_080722.1 | 3 | 303 | |||
GenBank mRNA | AF535869.1 | - | 3 | 653 | ||
AF535870.1 | - | 2 | 547 | |||
AK011740.2 | - | 3 | 252 | |||
AK019401.1 | - | 3 | 276 | |||
AK042531.1 | - | 3 | 251 | |||
AK154844.1 | - | 3 | 303 | |||
AY138505.1 | - | 2 | 149 | |||
BC003838.1 | - | 3 | 235 | 3490927 | ||
BG085760.2 | - | 3 | 454 | |||
BU694737.1 | - | 1 | 111 | |||
CF751080.1 | - | 2 | 217 |