Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291205.1 | NP_001278134.1 | 6 | 782 | ||
NM_001291206.1 | NP_001278135.1 | 6 | 1133 | |||
NM_001291207.1 | NP_001278136.1 | 5 | 678 | |||
NM_001291208.1 | NP_001278137.1 | 5 | 1027 | |||
NM_001291210.1 | NP_001278139.1 | 4 | 556 | |||
NM_026446.4 | NP_080722.1 | 6 | 784 | |||
GenBank mRNA | AF535869.1 | - | 6 | 1134 | ||
AF535870.1 | - | 5 | 1028 | |||
AK003967.1 | - | 2 | 200 | |||
AK011740.2 | - | 5 | 814 | |||
AK019401.1 | - | 6 | 757 | |||
AK042531.1 | - | 6 | 732 | |||
AK154844.1 | - | 6 | 784 | |||
AY138505.1 | - | 5 | 630 | |||
BC003838.1 | - | 6 | 716 | 3490927 |