Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047435.2 | NP_001040900.1 | 3 | 520 | ||
NM_001047436.2 | NP_001040901.1 | 3 | 536 | |||
NM_001286344.1 | NP_001273273.1 | 3 | 505 | |||
NM_001286345.1 | NP_001273274.1 | 3 | 433 | |||
NM_019695.3 | NP_062669.2 | 3 | 523 | |||
GenBank mRNA | AF070970.1 | - | 3 | 519 | ||
AK158848.1 | - | 3 | 518 | |||
BC049593.1 | - | 3 | 497 | 6771847 | ||
BC061114.1 | - | 3 | 433 | 6535523 | ||
BG806503.1 | - | 3 | 448 | |||
CK618363.1 | - | 3 | 479 | |||
CX206739.1 | - | 3 | 486 | |||
CX220562.1 | - | 3 | 530 |