Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019723.2 | NP_062697.1 | 9 | 1643 | ||
GenBank mRNA | AB018436.1 | - | 9 | 1643 | ||
AK080177.1 | - | 8 | 1846 | |||
AK133431.1 | - | 9 | 1572 | |||
AK161508.1 | - | 9 | 1539 | |||
BC132147.1 | - | 9 | 1606 | 40130424 | ||
BC137908.1 | - | 9 | 1606 | 8860927 |