Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019911.2 | NP_064295.2 | 1, 2 | 64 | ||
GenBank mRNA | AB476989.1 | - | 1, 2 | 538 | ||
AK132271.1 | - | 1, 2 | 87 | |||
BC018390.1 | - | 1, 2 | 64 | 4159877 | ||
U24493.1 | - | 1, 2 | 51 |