Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020046.3 | NP_064430.1 | 2 | 366 | ||
GenBank mRNA | AF029667.2 | - | 2 | 71 | ||
AK013102.1 | - | 2 | 318 | |||
AK088863.1 | - | 2 | 88 | |||
AK149350.1 | - | 2 | 114 | |||
BC019542.1 | - | 2 | 76 | 4039461 | ||
BC027829.1 | - | 2 | 327 | 5253723 | ||
BC045206.1 | - | 2 | 316 | 5253764 |