Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retnla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020509.3 | NP_065255.2 | 3 | 270 | ||
GenBank mRNA | AF205951.1 | - | 2 | 203 | ||
AF290872.1 | - | 3 | 308 | |||
AF316397.2 | - | 3 | 212 | |||
AF323082.1 | - | 3 | 280 | |||
AK007553.1 | - | 3 | 215 | |||
BC029248.1 | - | 2 | 210 | 4189285 |