Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tesc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021344.3 | NP_067319.2 | 6 | 542 | ||
GenBank mRNA | AF234783.1 | - | 6 | 448 | ||
AK010463.1 | - | 6 | 479 | |||
BC019492.1 | - | 6 | 542 | 4237590 | ||
BC107679.1 | - | 6 | 517 | 5290163 | ||
GQ856259.1 | - | 6 | 530 | |||
GQ856260.1 | - | 6 | 535 | |||
GQ856261.1 | - | 5 | 628 |