Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175098.5 | NP_780307.1 | 2 | 363 | ||
GenBank mRNA | AK018135.1 | - | 2 | 364 | ||
AK159264.1 | - | 2 | 393 | |||
BC095987.1 | - | 1 | 223 | 5365989 | ||
BC117837.2 | - | 2 | 311 | 40093139 |