Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021380.1 | NP_067355.1 | 3 | 326 | ||
GenBank mRNA | AF224267.1 | - | 3 | 326 | ||
AK078698.1 | - | 3 | 482 | |||
AY876923.1 | - | 2 | 190 | |||
BC111821.1 | - | 3 | 316 | 40049751 |