Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Repin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079901.1 | NP_001073370.2 | 3 | 702 | ||
NM_001079902.1 | NP_001073371.2 | 3 | 699 | |||
NM_001079903.1 | NP_001073372.2 | 3 | 631 | |||
NM_001079904.1 | NP_001073373.2 | 3 | 628 | |||
NM_001079905.1 | NP_001073374.1 | 2 | 513 | |||
NM_175099.3 | NP_780308.2 | 2 | 516 | |||
GenBank mRNA | AK089041.1 | - | 2 | 460 | ||
AK137310.1 | - | 3 | 575 | |||
AK170797.1 | - | 3 | 667 | |||
BC085128.1 | - | 3 | 634 | 5687175 | ||
BC092301.1 | - | 3 | 584 | |||
BM943840.1 | - | 2 | 474 |