Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159631.1 | NP_001153103.1 | 9, 10 | 953 | ||
NM_021606.3 | NP_067619.1 | 9, 10 | 946 | |||
GenBank mRNA | AF218847.1 | - | 9, 10 | 948 | ||
AK004925.1 | - | 9, 10 | 923 | |||
AK011519.1 | - | 9, 10 | 932 | |||
AK018168.1 | - | 9, 10 | 935 | |||
AK029266.1 | - | 9, 10 | 951 | |||
AK075717.1 | - | 9, 10 | 920 | |||
AK075836.1 | - | 9, 10 | 932 | |||
AK086700.1 | - | 951 | ||||
AK089919.1 | - | 9, 10 | 907 | |||
AK170757.1 | - | 9, 10 | 953 | |||
AK185671.1 | - | 2, 3 | 214 | |||
BC019524.1 | - | 9, 10 | 1015 | 4209097 |