Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021790.2 | NP_068562.1 | 3, 4 | 476 | ||
NM_181061.5 | NP_851406.1 | 3, 4 | 599 | |||
GenBank mRNA | AB043687.1 | - | 3, 4 | 476 | ||
AK082939.1 | - | 3, 4 | 256 | |||
AK160776.1 | - | 3, 4 | 234 | |||
BC028655.1 | - | 3, 4 | 599 | |||
BC092351.1 | - | 3, 4 | 238 |