Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021887.2 | NP_068687.1 | 5 | 863 | ||
GenBank mRNA | AB049137.1 | - | 5 | 755 | ||
AF254068.1 | - | 5 | 567 | |||
AF269134.1 | - | 4 | 425 | |||
AF279436.1 | - | 5 | 815 | |||
AF477982.1 | - | 4 | 441 | |||
AF477983.1 | - | 4 | 441 | |||
AF477984.1 | - | 4 | 441 | |||
AF477985.1 | - | 4 | 441 | |||
AF477986.1 | - | 4 | 441 | |||
AK040073.1 | - | 4 | 761 | |||
AK137793.1 | - | 4 | 1843 | |||
AK150824.1 | - | 5 | 824 | |||
AK171826.1 | - | 5 | 802 | |||
AK172032.1 | - | 5 | 835 | |||
BC057861.1 | - | 5 | 812 | 5315350 |